Learn More
Ansell™ Gladiator™ 16-650 Series Heavy Weight Gloves
Product Code. 11746168 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11746168 10 Each
12670772 8 12 pairs
12650772 10 12 pairs
12660772 7 12 pairs
12680772 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11746168 Supplier Ansell™ Supplier No. ACRZB1V08
missing translation for 'validMainframeMsg'
Designed for light-to-medium stress glass handling. Crinkle finish provides excellent wet and dry grip on slippery surfaces but also protects the hand from sharp edges.
  • Crinkle finish for grip on slipper surfaces and protection from sharp edges
  • Natural rubber coating
  • Cut-and-sewn construction
  • Interlock cotton liner
  • Fully-coated finish
  • Gauntlet cuff
Specifications
Coating Material Natural Rubber Latex
Color Green
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free No
Material Rubber Latex
Product Type Heavyweight Gloves
Size 10
Finger Style Crinkled finish
For Use With (Application) Glass repair and replacement Construction Utilities All food processing applications that have food contact Seafood processing
Glove Cuff Color Green
Glove Cuff Style Gauntlet
Liner Material Interlock Cotton
Lining Lined
Quantity 12 Pairs
Resistant To Good cushioning, good cut resistance
Series 16-650
Length (English) 12.2
Length (Metric) 310
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.