missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Glabridin

Product Code. 16458968
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458968 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458968 Supplier MedChemExpress Supplier No. HYN0393
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Glabridin is a natural isoflavan from Glycyrrhiza glabra, binds to and activates PPARγ, with an EC50 of 6115 nM. Glabridin exhibits antioxidant, anti-bacterial, anti-nephritic, anti-diabetic, anti-fungal, antitumor, anti-inflammatory, antiosteoporotic, cardiovascular protective, neuroprotective and radical scavenging activities.

Specifications

Content And Storage Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 324.37 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 59870-68-7
Product Type Natural Products
Purity 99.77%
Solubility DMSO : 25 mg/mL (77.07 mM∣Need ultrasonic)
Target PPAR∣Reactive Oxygen Species∣Bacterial
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.