Learn More
BOLA GL-Reductions
Click to view available options
Thread Size (Metric):
18
32
45
Unit Size:
Each
Description
- Black screw cap made of PPS, movable reduction body made of PTFE with O-ring made of FKM for transition to GL threads.
- The body can be moved independently from the screw cap so that the completely assembled reduction can be removed and fixed on another vessel without the risk of disarranging the tubing or cable.
- The product is only exposed to PTFE.
Specifications
Specifications
| Color | White |
| Length (Metric) | 0.8 to 8 mm |
| Sterility | Non-sterile |
| Thread Size (Metric) | 18 |
| Certifications/Compliance | FDA |
| Disposable | Reusable |
| Closure Liner | Unlined |
| Autoclavable | Autoclavable |