Learn More
BOLA GL-Reductions
Product Code. 15517029
Change view
Click to view available options
Thread Size (Metric):
18
32
45
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) unitSize
15517029 18 Each
15133603 32 Each
12843143 45 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15517029 Supplier BOLA Supplier No. D78401
Only null left
Add to basket
Add to basket
GL-Reductions PTFE/PPS
  • Black screw cap made of PPS, movable reduction body made of PTFE with O-ring made of FKM for transition to GL threads.
  • The body can be moved independently from the screw cap so that the completely assembled reduction can be removed and fixed on another vessel without the risk of disarranging the tubing or cable.
  • The product is only exposed to PTFE.
Specifications
Color White
Length (Metric) 0.8 to 8 mm
Sterility Non-sterile
Thread Size (Metric) 18
Certifications/Compliance FDA
Disposable Reusable
Closure Liner Unlined
Autoclavable Autoclavable