missing translation for 'onlineSavingsMsg'
Learn More

BOLA GL-Distributor with Stopcock

Product Code. 15650249
Click to view available options
Closure Size:
6x 14 - 1x 14
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15650249

Brand: BOLA E66114

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

GL-Distributor with Stopcock PTFE, 6 threaded necks GL 14 sideways

  • Distributor body made of PTFE with six lateral, GL-threaded connectors and one on the bottom.
  • Conical stopcock made of PTFE with L-Bore for good sealing and knurled grip made of PP for marking and regulation of flow direction.
  • Universal chemical resistance, the flowing product is only exposed to PTFE.

Specifications

Diameter (Metric) 7 x 8 mm
Autoclavable Autoclavable
Length (Metric) 80 mm
Closure Size 6x 14 - 1x 14
Certifications/Compliance FDA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.