Learn More
GKN1 Monoclonal Antibody (759821), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524367
Code : NEW
Additional Details : Weight : 0.00010kg
Description
This antibody detects mouse Gastrokine 1 in direct ELISAs and mouse and rat Gastrokine 1 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) Gastrokine 1 and no cross-reactivity with rhBlottin is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
GKN1 Monoclonal antibody specifically detects GKN1 in Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
GKN1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9CR36 | |
GKN1 | |
E. coli-derived recombinant mouse Gastrokine 1 Tyr38-Tyr201 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Immunohistochemistry (Frozen), Western Blot | |
759821 | |
Unconjugated | |
GKN1 | |
18 kDa antrum mucosa protein; 2200002K21Rik; Amp18; AMP-18; Amp18; gastrokine; BRICD1; BRICHOS domain containing 1; Ca11; FOV; foveolin; gastrokine 1; gastrokine-1; Gkn1; Protein CA11; Protein CA11 homolog; UNQ489/PRO1005 | |
Rat | |
Protein A/G | |
RUO | |
66283 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |