missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GKN1 Monoclonal Antibody (665701)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561660
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Gastrokine 1, also known as GKN1 and CA11, is a 185 amino acid containing secreted protein belonging to the gastrokine family with a BRICHOS domain and an asn-rich region. GKN1 (also known as an antioncogene) is reported to be down-regulated in human gastric cancer tissue as compared to normal gastric mucosa. Differential display of normal and cancerous gastric tissue suggested that this protein is predominantly expressed in stomach and the expression is down-regulated to considerable frequency in cancer tissue or gastric cancer cell lines examined. This extra cellular protein is co-secreted with mucins and is a signaling growth factor with mitogenic activity and may be involved in maintaining the integrity of the gastric mucosal epithelium.
Specifications
| GKN1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9NS71 | |
| GKN1 | |
| E. coli-derived recombinant human Gastrokine 1, Asn35-Asn199. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 665701 | |
| Unconjugated | |
| GKN1 | |
| 18 kDa antrum mucosa protein; 2200002K21Rik; Amp18; AMP-18; Amp18; gastrokine; BRICD1; BRICHOS domain containing 1; Ca11; FOV; foveolin; gastrokine 1; gastrokine-1; Gkn1; Protein CA11; Protein CA11 homolog; UNQ489/PRO1005 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 56287 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction