missing translation for 'onlineSavingsMsg'
Learn More

GKI-1, MedChemExpress

Product Code. p-200096850
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16493058

Brand: Medchem Express HY100521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GKI-1 is a Greatwall (GWL) kinase inhibitor with IC50s of 4.9 and 2.5 μM against hGWLFL and hGWL-KinDom, respectively. GKI-1 robustly inhibits ROCK1 with an IC50 of 11 μM, but only weakly affected PKA.

GKI-1 is a Greatwall (GWL) kinase inhibitor with IC50s of 4.9 and 2.5 μM against hGWLFL and hGWL-KinDom, respectively. GKI-1 robustly inhibits ROCK1 with an IC50 of 11 μM, but only weakly affected PKA.

Specifications

Chemical Name or Material GKI-1
Physical Form Solid
CAS 2444764-03-6
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H12ClN3
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 200 mg/mL (741.48 mM; Need ultrasonic)
SMILES ClC(C=C1)=CC=C1NC2=CC(C3=CNN=C3)=CC=C2
Molecular Weight (g/mol) 269.73
Formula Weight 269.73
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.