missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GKAP1, Mouse anti-Human, Clone: 1B9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00080318-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MASAVLSSVPTTASRFALLQVDSGSGSDSEPGKGKGRNTGKSQTLGSKSTTNEKKREKRRKKKEQQQSEANELRNLAFKKIPQKSSHAVCNAQHDLPLSNPVQKDSREENWQEWRQRDEQLTSEMFEADLEKALLLSKLEYEEHKKEYEDAENTSTQSKVMNKKDKRKNHQGKDRPLTVSLKDFHSEDHISKKTEELSSSQTLSHDGGFFNRLEDDVHKILIREKRREQLTEYNGTDNCTAHEHNQEVVLKDGRIERLKLELERKDAEIQKLKNVITQWEAKYKEVKARNAQLLKMLQEGEMKDKAEILLQVDESQSIKNELTIQVTSLHAALEQERSKVKVLQAELAKYQGGRKGKRNSESDQCRSpecifications
| GKAP1 | |
| Monoclonal | |
| Unconjugated | |
| G kinase anchoring protein 1 | |
| FKSG21/FLJ25469/GKAP42 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1B9 | |
| In 1x PBS, pH 7.4 | |
| BC014476 | |
| GKAP1 | |
| GKAP1 (AAH14476, 1 a.a. ∼ 366 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 80318 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu