missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GKAP/DLGAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98138-50ul
This item is not returnable.
View return policy
Description
GKAP/DLGAP1 Polyclonal antibody specifically detects GKAP/DLGAP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GKAP/DLGAP1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| DAP1, DAP-1-ALPHA, DAP-1DAP-1-BETA, discs, large (Drosophila) homolog-associated protein 1, disks large-associated protein 1, GKAPMGC88156, hGKAP, PSD-95/SAP90 binding protein 1, PSD-95/SAP90-binding protein 1, SAP90/PSD-95-associated protein 1, SAPAP1Guanylate kinase-associated protein | |
| Produced in rabbits immunized with E. coli-derived Human GKAP/DLGAP1 fragment. | |
| 50 μg | |
| Neuroscience | |
| 9229 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction