missing translation for 'onlineSavingsMsg'
Learn More

GKAP/DLGAP1 Antibody [Biotin], Novus Biologicals™

Product Code. 30734591 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734591 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734591 Supplier Novus Biologicals Supplier No. AF7296B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

GKAP/DLGAP1 Polyclonal antibody specifically detects GKAP/DLGAP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GKAP/DLGAP1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias DAP1, DAP-1-ALPHA, DAP-1DAP-1-BETA, discs, large (Drosophila) homolog-associated protein 1, disks large-associated protein 1, GKAPMGC88156, hGKAP, PSD-95/SAP90 binding protein 1, PSD-95/SAP90-binding protein 1, SAP90/PSD-95-associated protein 1, SAPAP1Guanylate kinase-associated protein
Host Species Sheep
Immunogen E. coli-derived recombinant human GKAP/DLGAP1, Lys368-Glu472, Accession # O14490
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9229
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.