missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GJC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04091-100UL
This item is not returnable.
View return policy
Description
GJC2 Polyclonal specifically detects GJC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GJC2 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57165 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| connexin46.6, connexin-46.6, Connexin-47, CX46.6, CX47, Gap junction alpha-12 protein, gap junction gamma-2 protein, gap junction protein, alpha 12, 47kDa, gap junction protein, gamma 2, 47kDa, GJA12connexin 47, HLD2, LMPH1C, PMLDAR, SPG44MGC105119 | |
| A synthetic peptide of human GJC2 | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction