missing translation for 'onlineSavingsMsg'
Learn More

GJC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55376-1-AP-150UL

 View more versions of this product

Product Code. 16866835

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

GJC2
Polyclonal
Unconjugated
GJC2
Connexin 46.6, Connexin 47, CX46.6, Cx47, Gap junction alpha 12 protein, Gap junction gamma 2 protein, GJA12, GJC2, HLD2, PMLDAR, SPG44
Rabbit
Antigen Affinity Chromatography
RUO
118454, 57165
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5T442, Q8BQU6
Gjc2
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.