Learn More
GJB5 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27166-1-AP-20UL
This item is not returnable.
View return policy
Description
The GJB5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GJB5. This antibody recognizes human, mouse antigen. The GJB5 antibody has been validated for the following applications: IP, ELISA analysis.
Specifications
| GJB5 | |
| 450 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunoprecipitation, ELISA | |
| Connexin 31.1, CX31.1, Gap junction beta 5 protein, GJB5 | |
| 20 μL | |
| 2709 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |