missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Givinostat hydrochloride
GREENER_CHOICE

Product Code. 18757946
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18757946

Brand: Tocris Bioscience™ 6249/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Histone deacetylase inhibitor

Givinostat hydrochloride is a histone deacetylase (HDAC) inhibitor (IC50 = 7.5 to 16 nM). At low concentrations reduces release of pro-inflammatory cytokines, including TNFα, INFγ, IL-1α and IL-1β, from LPS stimulated PBMCs in vitro. Exhibits anti-inflammatory effects in vivo. Improves functional recovery and attenuates tissue damage in a mouse model of traumatic brain injury.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [6-[(Diethylamino)methyl]-2-naphthalenyl]methyl-N-[4-[(hydroxyamino)carbonyl]phenyl]carbamate hydrochloride
CAS 199657-29-9
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C24H27N3O4.HCl
Formula Weight Observed MW: 457.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.