missing translation for 'onlineSavingsMsg'
Learn More

GITR/TNFRSF18 Antibody (YGITR765), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18406368 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406368 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406368 Supplier Novus Biologicals Supplier No. NBP281090MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GITR/TNFRSF18 Monoclonal antibody specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen GITR/TNFRSF18
Applications Flow Cytometry
Classification Monoclonal
Clone YGITR765
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18
Host Species Rabbit
Immunogen nB2/6TG cells transfected with mouse GITR/TNFRSF18 injected into DA rats.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8784
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.