missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR/TNFRSF18 Antibody (YGITR765), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81090F
This item is not returnable.
View return policy
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| GITR/TNFRSF18 | |
| Monoclonal | |
| FITC | |
| PBS | |
| TNFRSF18 | |
| nB2/6TG cells transfected with mouse GITR/TNFRSF18 injected into DA rats. | |
| 0.1 mL | |
| Apoptosis | |
| 8784 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| YGITR765 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody binds murine GITR/TNFRSF18. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction