Learn More
GITR/TNFRSF18 Antibody (CL7787) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21159AF532
Product Code. 30307203
  • £558.00 / 0.10mL
Description
GITR/TNFRSF18 Monoclonal antibody specifically detects GITR/TNFRSF18 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GITR/TNFRSF18
Monoclonal
Alexa Fluor 532
Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18
This antibody was developed using a synthetic peptide derived from Q9Y5U5, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis
8784
Store at 4⪚C in the dark.
IgG2a
Western Blot
CL7787
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified