missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR Ligand/TNFSF18 Antibody (YGL386), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81089DL594
This item is not returnable.
View return policy
Description
GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| GITR Ligand/TNFSF18 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| TNFSF18 | |
| Recombinant protein injected into DA rats. | |
| 0.1 mL | |
| Apoptosis | |
| 8995 | |
| Mouse | |
| Purified |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| YGL386 | |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody binds murine GITR Ligand/TNFSF18. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction