missing translation for 'onlineSavingsMsg'
Learn More

GITR Ligand/TNFSF18 Antibody (YGL386), Biotin, Novus Biologicals™

Product Code. 18379580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379580 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379580 Supplier Novus Biologicals Supplier No. NBP281089B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GITR Ligand/TNFSF18
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone YGL386
Conjugate Biotin
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18
Gene Symbols TNFSF18
Host Species Rabbit
Immunogen Recombinant protein injected into DA rats.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8995
Test Specificity This antibody binds murine GITR Ligand/TNFSF18.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.