missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR (CD357) Mouse anti-Human, BV421, Clone: V27-580, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566423
This item is not returnable.
View return policy
Description
GITR (CD357) Monoclonal antibody specifically detects GITR (CD357) in Human samples. It is validated for Flow Cytometry.
Specifications
| GITR (CD357) | |
| Monoclonal | |
| Brilliant Violet 421 | |
| 100 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2b κ |
| Flow Cytometry | |
| V27-580 | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction