missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590415
This item is not returnable.
View return policy
Description
Immunogen sequence: ASRLEKQNST PESDYDNTPN DMEPDGMGSS RKGRQRSMVW PGDGLVPDTA EPHVAPSPTL P; Positive Samples: A-431, LO2, Jurkat, HeLa.
GTPase-activating protein for the ADP ribosylation factor family.
Specifications
| GIT2 | |
| Polyclonal | |
| Unconjugated | |
| GIT2 | |
| 1500036H07Rik; 5830420E16Rik; 6430510B20Rik; 9630056M03Rik; ARF GAP GIT2; ARF GTPase activating protein 2; ARF GTPase-activating protein GIT2; B230104M05Rik; CAT2; CAT-2; cool associated tyrosine phosphorylated-2; cool-associated, tyrosine phosphorylated protein 2; cool-interacting tyrosine-phosphorylated protein 2; G protein-coupled receptor kinase interacting ArfGAP 2; G protein-coupled receptor kinase-interactor 2; GIT ArfGAP 2; Git2; GRK-interacting protein 2; KIAA0148; mKIAA0148; Paxillin kinase linker; PKL | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9815 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q14161 | |
| GIT2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-510 of human GIT2 (NP_4765111). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction