Learn More
GIT2 Monoclonal Antibody (M1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA520838
Description
Peptide Sequence: MSKRLRSSEV CADCSGPDPS WASVNRGTFL CDECCSVHRS LGRHISQVRH LKHTPWPPTL LQMVETLYNN GANSIWEHSL LDPASIMSGR RKANPQDKVH PNKAEFIRAK YQMLAFVHRL PCRDDDSVTA KDLSKQLHSS VRTGNLETCL RLLSLGAQAN FFHPEKGNTP LHVASKAGQI LQAELLAVYG ADPGTQDSSG KTPVDYARQG GHHELAERLV EIQYELTDRL AFYLCGRKPD HKNGQHFIIP QMADSSLDLS ELAKAAKKKL QSLSNHLFEE LAMDMYDEVD RRETDAVWLA TQNHSALVTE TTVVPFLPVN PEYSSTRNQG RQKLARFNAH EFATLVIDIL SDAKRRQQGS SLSGSKDNVE LILKTINNQH SVESQDNDQP DYDSVASDED TDLETTASKT NRQKSLDSDL SDGPVTVQEF MEVKNALVAS EAKIQQLMKV NNNLSDELRI MQKKLLGKDA N
This gene encodes a member of the GIT protein family, which interact with G protein-coupled receptor kinases and possess ADP-ribosylation factor (ARF) GTPase-activating protein (GAP) activity. GIT proteins traffic between cytoplasmic complexes, focal adhesions, and the cell periphery, and interact with Pak interacting exchange factor beta (PIX) to form large oligomeric complexes that transiently recruit other proteins. GIT proteins regulate cytoskeletal dynamics and participate in receptor internalization and membrane trafficking. This gene has been shown to repress lamellipodial extension and focal adhesion turnover, and is thought to regulate cell motility. This gene undergoes extensive alternative splicing to generate multiple isoforms, but the full-length nature of some of these variants has not been determined. The various isoforms have functional differences, with respect to ARF GAP activity and to G protein-coupled receptor kinase 2 binding.
Specifications
| GIT2 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q14161 | |
| GIT2 | |
| GIT2 (AAH01379, 1 a.a. ∽ 471 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| M1 | |
| Unconjugated | |
| GIT2 | |
| CAT-2, CAT2, ARF GAP GIT2, ARF GTPase-activating protein GIT2, G protein-coupled receptor kinase-interactor 2, GRK-interacting protein 2, cool-associated, tyrosine phosphorylated protein 2, cool-interacting tyrosine-phosphorylated protein 2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9815 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.