missing translation for 'onlineSavingsMsg'
Learn More

GIT2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18421443 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421443 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421443 Supplier Novus Biologicals Supplier No. NBP119143AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GIT2 Polyclonal antibody specifically detects GIT2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIT2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ARF GAP GIT2, ARF GTPase-activating protein GIT2, CAT2, CAT-2, cool-associated, tyrosine phosphorylated protein 2, Cool-interacting tyrosine-phosphorylated protein 2, DKFZp686G01261, G protein-coupled receptor kinase interacting ArfGAP 2, G protein-coupled receptor kinase interactor 2, G protein-coupled receptor kinase-interactor 2, KIAA0148GRK-interacting protein 2, MGC760
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 500 and 550 of human G protein-coupled receptor kinase interacting ArfGAP 2 using the numbering given in entry NP_476510.1 (GeneID 9815).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9815
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.