missing translation for 'onlineSavingsMsg'
Learn More

GIT2 Antibody (1B2), Novus Biologicals™

Product Code. 18396179 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396179 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396179 Supplier Novus Biologicals Supplier No. H00009815M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GIT2 Monoclonal antibody specifically detects GIT2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIT2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1B2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055591
Gene Alias ARF GAP GIT2, ARF GTPase-activating protein GIT2, CAT2, CAT-2, cool-associated, tyrosine phosphorylated protein 2, Cool-interacting tyrosine-phosphorylated protein 2, DKFZp686G01261, G protein-coupled receptor kinase interacting ArfGAP 2, G protein-coupled receptor kinase interactor 2, G protein-coupled receptor kinase-interactor 2, KIAA0148GRK-interacting protein 2, MGC760
Host Species Mouse
Immunogen GIT2 (NP_055591, 401 a.a. ~ 498 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TDLETTASKTNRQKLQTLQSENSNLRKQATTNVYQVQTGSEYTDTSNHSSLKRRPSARGSRPMSMYETGSGQKPYLPMGEASRPEESRMRLQPFPAHA
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9815
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.