missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Git1, Mouse anti-Rat, ATTO488, Clone: S39B-8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB18347.100ug
This item is not returnable.
View return policy
Specifications
| Git1 | |
| Monoclonal | |
| Atto 488 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
| - | |
| Mouse | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| S39B-8 | |
| Immunocytochemistry Immunofluorescence Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| G protein-coupled receptor kinase interacting ArfGAP 1 | |
| Git1 | |
| Recombinant protein corresponding to amino acids 375-770 at C-terminus of rat Git1. | |
| RUO | |
| 83709 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction