missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIT1 Antibody (S39B-8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22423H
This item is not returnable.
View return policy
Description
GIT1 Monoclonal specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Knockout Validated.
Specifications
| GIT1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| GIT1 | |
| Fusion protein amino acids 375-770 (C-terminus) of rat GIT1 | |
| 0.1 mL | |
| 28964 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoassay | |
| S39B-8 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Knockout Validated | |
| ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects approx 90kDa. Does not cross-react with GIT2. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction