missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Gisadenafil besylate
GREENER_CHOICE

Product Code. p-4816686
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18006032 10 mg 10mg
18065542 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18006032 Supplier Tocris Bioscience™ Supplier No. 4165/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent PDE5 inhibitor; orally bioavailable

Potent PDE5 inhibitor (IC50 = 1.23 nM). Shows >100-fold selectivity for PDE5 over PDE6. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334827-98-4
Molecular Formula C29H39N7O8S2

Specifications

Quantity 10 mg
Formula Weight 677.79
Purity >99%
Molecular Weight (g/mol) 677.79
Chemical Name or Material Gisadenafil besylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.