missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIRK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21647-1-AP-20UL
This item is not returnable.
View return policy
Description
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins and may be involved in the regulation of insulin secretion by glucose. It associates with two other G-protein-activated potassium channels to form a heteromultimeric pore-forming complex.Specifications
| GIRK2 | |
| Polyclonal | |
| Unconjugated | |
| KCNJ6 | |
| BIR1, GIRK 2, GIRK2, hiGIRK2, KATP 2, KATP2, KCNJ6, KCNJ7, KIR3.2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16522, 25743, 3763 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P48051, P48542, P48550 | |
| KCNJ6 | |
| GIRK2 Fusion Protein Ag16344 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction