missing translation for 'onlineSavingsMsg'
Learn More

Girdin Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18409044 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409044 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409044 Supplier Novus Biologicals Supplier No. NBP171857AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Girdin Polyclonal antibody specifically detects Girdin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Girdin
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Akt phosphorylation enhancer, APEGIRDIN, coiled-coil domain containing 88A, Coiled-coil domain-containing protein 88A, G alpha-interacting vesicle-associated protein, G{alpha}-interacting vesicle-associated protein, Girders of actin filament, girdin, GIVFLJ10392, GRDNAKT-phosphorylation enhancer, HKRP1, HkRP1Galpha-interacting vesicle-associated protein, Hook-related protein 1, KIAA1212DKFZp686D0630
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1425 and 1475 of human Coiled-Coil Domain Containing 88A using the numbering given in entry NP_001129069.1 (GeneID 55704).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 55704
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.