missing translation for 'onlineSavingsMsg'
Learn More

Girard's Reagent T, TRC

Product Code. 30393656
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393656 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393656 Supplier Toronto Research Chemicals Supplier No. TRCG388505250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Girard's Reagent T
CAS 123-46-6
Molecular Formula C5H14ClN3O
Quantity 250 g
Synonym 2-Hydrazinyl-N,N,N-trimethyl-2-oxo-ethanaminium Chloride, (Carboxymethyl)trimethylammonium Chloride Hydrazide, (Carboxymethyl)trimethyl-ammonium Chloride Hydrazide, 2-Hydrazino-N,N,N-trimethyl-2-oxo-ethanaminium Chloride, (2-Hydrazino-2-oxoethyl)trimethylammonium Chloride, (Carbazoylmethyl)trimethylammonium Chloride, Betaine Hydrazide Chloride, Betaine Hydrazide Hydrochloride, Girard-T Reagent, Girard's T Reagent, Girard's T Reagents, Trimethylacetylhydrazide Ammonium Chloride, Trimethylaminoacetohydrazide Chloride, Trimethylammonioacetohydrazide Chloride, Trimethylammonium Acetyl Hydrazide Chloride, Trimethylammonium Chloride Acetylhydrazide
InChI Formula InChI=1S/C5H13N3O.ClH/c1-8(2,3)4-5(9)7-6;/h4,6H2,1-3H3;1H
SMILES [Cl-].C[N+](C)(C)CC(=O)NN
IUPAC Name (2-hydrazinyl-2-oxoethyl)-trimethylazanium;chloride
Molecular Weight (g/mol) 167.64
Formula Weight 167.0825
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.