missing translation for 'onlineSavingsMsg'
Learn More

Girard’s Reagent P-d5, TRC

Product Code. 30393294
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393294 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30393294 Supplier Toronto Research Chemicals Supplier No. TRCG3885021G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Girard's Reagent P-d5
CAS 1505505-87-2
Purity Grade Notes HPLC
Molecular Formula C7H5D5ClN3O
Quantity 1 g
Synonym 1-(Carboxymethyl)pyridinium-d6 Chloride Hydrazide, 1-(2-Hydrazino-2-oxoethyl)pyridinium-d5 Chloride, Girard-P Reagent-d5, Girard’s P Reagent-d5, (Pyridinio-d5)acetohydrazide Chloride
InChI Formula InChI=1S/C7H9N3O.ClH/c8-9-7(11)6-10-4-2-1-3-5-10;/h1-5H,6,8H2;1H/i1D,2D,3D,4D,5D;
SMILES NNC(C[N+]1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])=O.[Cl-]
IUPAC Name 2-(2,3,4,5,6-pentadeuteriopyridin-1-ium-1-yl)acetohydrazide;chloride
Molecular Weight (g/mol) 192.66
Formula Weight 192.0826
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.