missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GIPR Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA533586

Product Code. 15324521

  • £442.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BLAST analysis of the peptide immunogen showed 100% homology with human protein GHRHR. Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%) Marmoset (95%) Dog, Hamster, Panda (89%) Mouse, Rat, Rabbit (84%).

GIPR encodes a G-protein coupled receptor for gastric inhibitory polypeptide (GIP), which was originally identified as an activity in gut extracts that inhibited gastric acid secretion and gastrin release, but subsequently was demonstrated to stimulate insulin release in the presence of elevated glucose. Mice lacking this gene exhibit higher blood glucose levels with impaired initial insulin response after oral glucose load. Defect in this gene thus may contribute to the pathogenesis of diabetes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GIPR
Polyclonal
Unconjugated
GIPR
gastric inhibitory peptide receptor; gastric inhibitory polypeptide receptor; GIP receptor; Gippr; Gipr; GIP-R; Glucose-dependent insulinotropic polypeptide receptor; Gm1081; Gm160; PGQTL2; RATGIPPR
Rabbit
Antigen affinity chromatography
RUO
2696
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
P48546
GIPR
Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GIPR.
50 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.