missing translation for 'onlineSavingsMsg'
Learn More

GIPR, Mouse anti-Human,Mouse,Rat,Canine, Clone: Gipg013, Novus Biologicals™

Product Code. 18398910 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398910 0.2 mg 0.2mg
18328190 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18398910 Supplier Novus Biologicals Supplier No. NBP2810670.2MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GIPR Monoclonal specifically detects GIPR in Human, Mouse, Rat, Canine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIPR
Applications ELISA, Immunofluorescence, Blocking Assay
Classification Monoclonal
Clone Gipg013
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize
Formulation PBS
Gene Alias gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2
Gene Symbols GIPR
Host Species Mouse
Immunogen Human GIPR.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2696
Test Specificity The original has a Ki value of 7 nM and a Kd value of 6.8 nM for human GIPR.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.