missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIPR, Mouse anti-Human,Mouse,Rat,Canine, Clone: Gipg013, Novus Biologicals™
Mouse Monoclonal Antibody
£217.00 - £473.00
Specifications
| Antigen | GIPR |
|---|---|
| Clone | Gipg013 |
| Concentration | 1 mg/mL |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize |
| Applications | ELISA, Immunofluorescence, Blocking Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18328190
|
Novus Biologicals
NBP2-81067-0.025MG |
0.025 mg |
£217.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18398910
|
Novus Biologicals
NBP2-81067-0.2MG |
0.2 mg |
£473.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GIPR Monoclonal specifically detects GIPR in Human, Mouse, Rat, Canine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize.Specifications
| GIPR | |
| 1 mg/mL | |
| ELISA, Immunofluorescence, Blocking Assay | |
| Unconjugated | |
| Mouse | |
| GPCR, Neuroscience | |
| PBS | |
| 2696 | |
| Human GIPR. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Gipg013 | |
| ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Canine | |
| gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2 | |
| GIPR | |
| IgG2a λ | |
| Protein A purified | |
| The original has a Ki value of 7 nM and a Kd value of 6.8 nM for human GIPR. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel