missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIPR (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577447
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
This gene encodes a G-protein coupled receptor for gastric inhibitory polypeptide (GIP), which was originally identified as an activity in gut extracts that inhibited gastric acid secretion and gastrin release, but subsequently was demonstrated to stimulate insulin release in the presence of elevated glucose. Mice lacking this gene exhibit higher blood glucose levels with impaired initial insulin response after oral glucose load. Defect in this gene thus may contribute to the pathogenesis of diabetes.
Specifications
| GIPR (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| Gipr | |
| Gastric Inhibitory Polypeptide Receptor (extracellular) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25024, 381853 | |
| -20°C | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P43219, Q0P543 | |
| Gipr | |
| Peptide (C)EGQTTGELYQRWER, corresponding to amino acid residues 25-38 of rat Gastric Inhibitory Polypeptide Receptor | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction