missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GIPR Monoclonal antibody specifically detects GIPR in Human, Mouse, Rat, Canine samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Neutralization
Specifications
Specifications
| Antigen | GIPR |
| Applications | ELISA, Neutralization |
| Immunogen | Human GIPR. |
| Gene ID (Entrez) | 2696 |
| Target Species | Rat, Canine |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?