Learn More
GIPR Antibody (Gipg013), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-81067F
Product Code. 18305531
  • £496.00 / 0.10mL
Description
GIPR Monoclonal specifically detects GIPR in Human, Mouse, Rat, Canine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GIPR
Monoclonal
FITC
PBS
GIPR
Human GIPR.
0.1 mL
GPCR, Neuroscience
2696
Human, Mouse, Rat, Canine
Purified
ELISA, Immunofluorescence
Gipg013
ELISA, Immunocytochemistry/Immunofluorescence
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2
Mouse
Protein A purified
RUO
Primary
The original has a Ki value of 7 nM and a Kd value of 6.8 nM for human GIPR.
Store at 4°C in the dark.
IgG2a λ