missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GIPR Monoclonal specifically detects GIPR in Human, Mouse, Rat, Canine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | GIPR |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | Gipg013 |
| Conjugate | DyLight 755 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2 |
| Gene Symbols | GIPR |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?