missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GIPC3 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA518227

Product Code. 11396193

  • £442.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Required for postnatal maturation of the hair bundle and long-term survival of hair cells and spiral ganglion.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GIPC3
Polyclonal
Unconjugated
GIPC3
C19orf64; DFNB15; DFNB72; DFNB95; DKFZp686J1198; FLJ40925; GIPC PDZ domain containing family member 3; GIPC PDZ domain containing family, member 3; GIPC3; PDZ domain-containing protein GIPC3
Goat
Ammonium sulfate precipitation
RUO
126326
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q8TF64
GIPC3
Synthetic peptide sequence (EGAAAREARGTET) corresponding to the N-terminus amino acids of GIPC3 (aa 2-15).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.