missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIPC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31178
This item is not returnable.
View return policy
Description
GIPC3 Polyclonal specifically detects GIPC3 in Human samples. It is validated for Western Blot.
Specifications
| GIPC3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| GIPC3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human GIPC3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 126326 | |
| Human | |
| IgG |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| C19orf64, chromosome 19 open reading frame 64, DKFZp686J1198, FLJ40925, GIPC PDZ domain containing family, member 3, PDZ domain protein GIPC3, PDZ domain-containing protein GIPC3 | |
| Rabbit | |
| 34 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction