missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GIPC2 Antibody, Novus Biologicals™

Product Code. 18344989 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344989 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344989 Supplier Novus Biologicals™ Supplier No. H00054810B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

GIPC2 Polyclonal antibody specifically detects GIPC2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen GIPC2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS (pH 7.4)
Gene Accession No. AAH36075.1
Gene Alias FLJ20075, GIPC PDZ domain containing family, member 2, PDZ domain protein GIPC2, PDZ domain-containing protein GIPC2, semaF cytoplasmic domain associated protein 2, semaphorin cytoplasmic domain associated protein 2, SEMCAP2, SEMCAP-2
Host Species Mouse
Immunogen GIPC2 (AAH36075.1, 1 a.a. - 315 a.a.) full-length human protein. MPLKLRGKKKAKSKETAGLVEGEPTGAGGGSLSASRAPARRLVFHAQLAHGSATGRVEGFSSIQELYAQIAGAFEISPSEILYCTLNTPKIDMERLLGGQLGLEDFIFAHVKGIEKEVNVYKSEDSLGLTITDNGVGYAFIKRIKDGGVIDSVKTICVGDHIESINGENIVGWRHYDVAKKLKELKKEELFTMKLIEPKKAFEIEPRSKAGKSSGEKIGCGRATLRLRSKGPATVEEMPSETKAKAIEKIDDVLELYMGIRDIDLATTMFEAGKDKVNPDEFAVALDETLGDFAFPDEFVFDVWGVIGDAKRRGL
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54810
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.