missing translation for 'onlineSavingsMsg'
Learn More

GIPC2 Antibody (1G10), DyLight 650, Novus Biologicals™

Product Code. 18400317 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400317 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400317 Supplier Novus Biologicals Supplier No. NBP222552C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GIPC2 Monoclonal antibody specifically detects GIPC2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIPC2
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1G10
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias FLJ20075, GIPC PDZ domain containing family, member 2, PDZ domain protein GIPC2, PDZ domain-containing protein GIPC2, semaF cytoplasmic domain associated protein 2, semaphorin cytoplasmic domain associated protein 2, SEMCAP2, SEMCAP-2
Host Species Mouse
Immunogen Recombinant human GIPC2 (1-315aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54810
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.