missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIPC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583696
This item is not returnable.
View return policy
Description
Immunogen sequence: EAINGQSLLG CRHYEVARLL KELPRGRTFT LKLTEPRKAF DMISQRSAGG RPGSGPQLGT GR Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 98%.
GIPC1 is a scaffolding protein that regulates cell surface receptor expression and trafficking (Lee et al., 2008).
Specifications
| GIPC1 | |
| Polyclonal | |
| Unconjugated | |
| GIPC1 | |
| C19orf3; GAIP C-terminus-interacting protein; Gipc; GIPC PDZ domain containing family member 1; GIPC PDZ domain containing family, member 1; Gipc1; GLUT1 C-terminal binding protein; GLUT1 C-terminal-binding protein; GLUT1CBP; Glut1CIP; Hs.6454; IGF-1 receptor interacting protein 1; IIP-1; neurophilin1-IP; NIP; PDZ domain-containing protein GIPC1; regulator of G-protein signaling 19 interacting protein 1; regulator of G-protein signalling 19 interacting protein 1; Rgs19; RGS19-interacting protein 1; RGS19IP1; RGS-GAIP interacting protein C-terminus; RGS-GAIP interacting protein GIPC; RGS-GAIP interacting protein, C-terminus; RGS-GAIP-interacting protein; semaF cytoplasmic domain associated protein; semaF cytoplasmic domain-associated protein 1; SEMCAP; Semcap1; SEMCAP-1; SYNECTIIN; SYNECTIN; Tax interacting protein, clone 2; tax interaction protein 2; TaxIP2; TIP-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10755 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14908 | |
| GIPC1 | |
| Recombinant protein corresponding to Human GIPC1. Recombinant protein control fragment (Product #RP-99024). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion