missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIPC1 Monoclonal Antibody (24GB6645)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557415
This item is not returnable.
View return policy
Description
GIPC1 Monoclonal Antibody for Western Blot
GIPC1 is a scaffolding protein that regulates cell surface receptor expression and trafficking (Lee et al., 2008).
Specifications
| GIPC1 | |
| Monoclonal | |
| Unconjugated | |
| GIPC1 | |
| C19orf3; GAIP C-terminus-interacting protein; Gipc; GIPC PDZ domain containing family member 1; GIPC PDZ domain containing family, member 1; Gipc1; GLUT1 C-terminal binding protein; GLUT1 C-terminal-binding protein; GLUT1CBP; Glut1CIP; Hs.6454; IGF-1 receptor interacting protein 1; IIP-1; neurophilin1-IP; NIP; PDZ domain-containing protein GIPC1; regulator of G-protein signaling 19 interacting protein 1; regulator of G-protein signalling 19 interacting protein 1; Rgs19; RGS19-interacting protein 1; RGS19IP1; RGS-GAIP interacting protein C-terminus; RGS-GAIP interacting protein GIPC; RGS-GAIP interacting protein, C-terminus; RGS-GAIP-interacting protein; semaF cytoplasmic domain associated protein; semaF cytoplasmic domain-associated protein 1; SEMCAP; Semcap1; SEMCAP-1; SYNECTIIN; SYNECTIN; Tax interacting protein, clone 2; tax interaction protein 2; TaxIP2; TIP-2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10755, 67903, 83823 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB6645 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14908, Q9Z0G0, Q9Z254 | |
| GIPC1 | |
| Recombinant protein of human GIPC1 (aa 1-333). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction