missing translation for 'onlineSavingsMsg'
Learn More

GIPC1 Antibody, Novus Biologicals™

Product Code. 18612831 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
25 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18612831 25 μL 25µL
18386751 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18612831 Supplier Novus Biologicals Supplier No. NBP27655725ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GIPC1 Polyclonal specifically detects GIPC1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIPC1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 u/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
Gene Alias C19orf3chromosome 19 open reading frame 3, GAIP C-terminus-interacting protein, GIPC PDZ domain containing family, member 1, GIPCRGS-GAIP-interacting protein, GLUT1 C-terminal binding protein, GLUT1CBP, IGF-1 receptor interacting protein 1, MGC15889, MGC3774, NIP, PDZ domain-containing protein GIPC1, regulator of G-protein signalling 19 interacting protein 1, RGS19IP1IIP-1, SEMCAP, SYNECTIIN, SYNECTIN, Tax interaction protein 2, TIP-2RGS19-interacting protein 1
Gene Symbols GIPC1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: MPLGLGRRKKAPPLVENEEAEPGRG
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10755
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.