missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £393.00
Specifications
| Antigen | GIP |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18317904
|
Novus Biologicals
NBP3-04865-20UL |
20 μL |
£171.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18340517
|
Novus Biologicals
NBP3-04865-100UL |
100 μL |
£393.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GIP Polyclonal specifically detects GIP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GIP | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 2695 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-153 of human GIP (NP_004114.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title