Learn More
Invitrogen™ GIP Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA579310
Product Code. 15914885
  • £392.00 / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, human HepG2 whole cell. IHC: human colorectal adenocarcinoma tissue, rat intestine tissue.
This gene encodes an incretin hormone and belongs to the glucagon superfamily. The encoded protein is important in maintaining glucose homeostasis as it is a potent stimulator of insulin secretion from pancreatic beta-cells following food ingestion and nutrient absorption. This gene stimulates insulin secretion via its G protein-coupled receptor activation of adenylyl cyclase and other signal transduction pathways. It is a relatively poor inhibitor of gastric acid secretion.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GIP
Polyclonal
Unconjugated
Gip
gastric inhibitory polypeptide; Gip; Glucose-dependent insulinotropic peptide; glucose-dependent insulinotropic polypeptide; Gludins; incretin hormone; Rabbit Anti GIP; RATGLUDINS
Rabbit
Antigen affinity chromatography
RUO
25040, 2695
-20°C
Lyophilized
Immunohistochemistry (Paraffin), Western Blot
500 μg/mL
Antibody with 0.2mg Na2PO4, 0.9mg NaCl, 4mg trehalose and 0.01mg sodium azide
P09681, Q06145
Gip
E. coli-derived human GIP recombinant protein (Position: Y52-Q93).
100 μg
Primary
Human, Rat