missing translation for 'onlineSavingsMsg'
Learn More

GIP Antibody (14), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18386683 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386683 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386683 Supplier Novus Biologicals Supplier No. NBP312092AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GIP Monoclonal antibody specifically detects GIP in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIP
Applications ELISA
Classification Monoclonal
Clone 14
Conjugate Alexa Fluor 350
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide
Host Species Mouse
Immunogen Synthetic human gastric inhibitory polypeptide
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2695
Target Species Human
Content And Storage Store at 4°C in the dark.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.