missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside Rg1, TRC
Specifications
Specifications
| Chemical Name or Material | Ginsenoside Rg1 |
| CAS | 22427-39-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C42 H72 O14 |
| Quantity | 250 mg |
| Synonym | Ginsenoside Rg1, Panaxoside A, Dammarane, β-D-glucopyranoside deriv., Ginsenoside A2, Ginsenoside g1, Panaxoside Rg1, Panaxsaponin Rg1, Sanchinoside C1, Sanchinoside Rg1 |
| InChI Formula | InChI=1S/C42H72O14/c1-20(2)10-9-13-42(8,56-37-34(52)32(50)30(48)25(19-44)55-37)21-11-15-40(6)28(21)22(45)16-26-39(5)14-12-27(46)38(3,4)35(39)23(17-41(26,40)7)53-36-33(51)31(49)29(47)24(18-43)54-36/h10,21-37,43-52H,9,11-19H2,1-8H3/t21-,22+,23-,24+,25+,26+,27-,28-,29+,30+,31-,32-,33+,34+,35-,36+,37-,39+,40+,41+,42-/m0/s1 |
| SMILES | CC(=CCC[C@](C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H]2CC[C@]3(C)[C@@H]2[C@H](O)C[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5[C@H](C[C@@]34C)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-[[(3S,5R,6S,8R,9R,10R,12R,13R,14R,17S)-3,12-dihydroxy-4,4,8,10,14-pentamethyl-17-[(2S)-6-methyl-2-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhept-5-en-2-yl]-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-6-yl]oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 801.01 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?